Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LPA (Human) ELISA Kit

Catalog No. 89028151 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-151 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-151 Supplier Abnova™ Supplier No. KA1041
Add to Cart
Add to Cart

Includes

  • A 96-well polystyrene Human Lp(a) microplate (12 strips of 8 wells) coated with a polyclonal antibody against human Lp(a), 1 in no.
  • Pressure-sensitive sealing tapes, which can be cut to fit the format of the individual assay, 3 in nos.
  • Human Lp(a) standard in a buffered protein base (lyophilized), 50ng
  • Biotinylated Lp(a) Antibody: A 50-fold concentrated biotinylated polyclonal antibody against Lp(a), 140μL
  • EIA Diluent Concentrate: A 10-fold concentrated buffered protein base 30mL
  • A 20-fold concentrated wash buffered surfactant 30mL, 2 bottles
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate 80μL
  • A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine, 8mL
  • Stop Solution: A 0.5N hydroxychloric acid to stop the chromogen substrate reaction, 12mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Human LPA in Plasma, Serum, Urine, Milk and cell culture samples

Specifications

Calibration Range 0.78 to 50ng/mL
Conjugate HRP
Cross Reactivity Human
Description LPA (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human LPA in plasma, serum, urine, milk, and cell culture samples.
Detection Limit 0.7ng/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 50ng Human LPA standard, 1 vial 50X Biotinylated LPA Antibody, 30mL 10X EIA Diluent concentrate, 2 x 30mL Wash Buffer 20X concentrate, 80μL Streptavidin-Peroxidase Conjugate (SP Conjugate),, 8mL chromog
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Milk, Plasma, Serum, Urine
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type LPA (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.