Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LTF (Chicken) ELISA Kit

Catalog No. 89100356 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-356 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-356 Supplier Abnova™ Supplier No. KA2124
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Diluent Concentrate (Running Buffer)
  • Wash Solution Concentrate
  • Enzyme-Antibody Conjugate: 100X concentrated affinity-purified anti-Chicken Ovotransferrin antibody conjugated with HRP in stabilizing buffer
  • Chromogenic Substrate Solution: 3,3',5,5'-tetramethybenzidine (TMB) and hydrogen peroxide in citric acid buffer at pH 3.3
  • Stop Solution: 0.3M sulfuric acid. WARNING: Avoid contact with skin
  • Anti-Chicken Ovotransferrin ELISA micro plate
  • Chicken Ovotransferrin Calibrator

Measure chicken LTF. Two-site enzyme linked immunoassay.

The LTF (Chicken) ELISA Kit is a two-site enzyme linked immunoassay (ELISA) for measuring Ovotransferrin in biological samples of Chickens

Specifications

Conjugate HRP
Cross Reactivity Chicken
Description LTF (Chicken) ELISA Kit is a two-site enzyme linked immunoassay for the measurement of chicken LTF.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Biological Fluid
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the calibrator at -20°C for long term storage. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.