Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LTF (Human) ELISA Kit

Catalog No. 89022911 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-911 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-911 Supplier Abnova™ Supplier No. KA0484
Add to Cart
Add to Cart

Includes

  • Human Lactoferrin Microplate: A polystyrene microplate coated with a polyclonal antibody against human Lactoferrin, 96-well
  • Pressure-sensitive sealing tapes that can be cut to fit the format of the individual assay, 3 in no.
  • Human Lactoferrin in a buffered protein base, lyophilized, 640ng
  • 100-fold concentrated biotinylated polyclonal antibody against human lactoferrin, 80μL
  • 10-fold MIX diluent concentrated buffered protein base, 30mL
  • 20-fold concentrated wash buffered surfactant, 30mL x 2
  • 100-fold concentrate Streptavidin-Peroxidase Conjugate (SP Conjugate), 80μL
  • Stabilized peroxidase chromogen substrate tetramethylbenzidine, Ready to use, 8mL
  • 0.5 N hydroxychloric acid to stop the chromogen substrate reaction, 12mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Human LTF

Specifications

Calibration Range 0.625 to 40ng/mL
Conjugate Biotin
Cross Reactivity Human
Description LTF (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human LTF.
Detection Limit 0.6ng/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 640ng Human Lactoferrin standard, 1 vial 100X Biotinylated Lactoferrin Antibody, 30mL 10X MIX Diluent concentrate, 2 x 30mL Wash Buffer 20X concentrate, 80 Streptavidin-Peroxidase Conjugate (SP Conjugate),, 8mL Chromogen Substrate, 12mL stop solution
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Cerebrospinal Fluid, Milk, Plasma, Saliva, Serum, Urine
Sample Volume 50 μL
Content And Storage Store SP Conjugate and Biotinylated Antibody at -20°C. Store Standard at 4°C before reconstituting with Diluent and at -20°C after reconstituting with Diluent. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type LTF (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.