Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Luciferase Reporter Gene Assay Kit

Catalog No. 89028372 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-372 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-372 Supplier Abnova™ Supplier No. KA1640
Add to Cart
Add to Cart

Bioluminescent assay for promoter regulated luciferase expression

Specifications

Product Type Reporter Gene Assay Kit
Description Luciferase Reporter Gene Assay Kit is a bioluminescent assay for promoter regulated luciferase expression.
Includes 20mL assay buffer, reagent 1 each
Content And Storage Store Reagent at -20°C. Store Assay Buffer at 4°C.
For Use With (Application) Functional Study
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.