Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MAP2K1 (pS217/221)/Pan MAP2K (Human/Mouse/Rat) ELISA Kit

Catalog No. 89100446 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-446 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-446 Supplier Abnova™ Supplier No. KA2166

Includes

  • Mek1 Microplate (Item A): Coated with anti-pan-Mek1 antibody
  • Wash Buffer Concentrate (20x) (Item B)
  • Assay Diluent (Item E)
  • Detection Antibody Mek1 (Ser217/221) (Item C-1): Rabbit anti-phosphoMek1 (Ser217/221)
  • Detection Antibody Mek1 (Item C-2): Rabbit anti-pan-Mek1
  • HRP-conjugated Anti-rabbit IgG (Item D-1): 500x HRP-conjugated Anti-rabbit IgG concentrate, 25μL
  • TMB One-Step Substrate Reagent (Item H)
  • Stop Solution (Item I): 0.2M sulfuric acid
  • Cell Lysate Buffer (Item J)
  • Positive Control HelaT003-1 (Item K): lyophilized powder from treated Hela cell lysate
  • Sealing Tape

Use to detect qualitatively human, mouse, rat phospho MAP2K1 (S217/221) and Pan MAP2K. Enzyme-linked immunosorbent assay.

This Sandwich ELISA kit is an in vitro enzyme-linked immunosorbent assay for the measurement of phospho-Mek1 (Ser217/221) and pan Mek1 in human, mouse and rat cell lysates

Specifications

Conjugate HRP
Cross Reactivity Human, Mouse, Rat
Description MAP2K1 (pS217/221)/Pan MAP2K (Human/Mouse/Rat) ELISA Kit is an enzyme-linked immunosorbent assay for the qualitative determination of human, mouse, rat phospho MAP2K1 (S217/221) and Pan MAP2K.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Cell Lysate, Tissue Lysate
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Qualitative
Product Type MAP2K1 (pS217/221)/Pan MAP2K (Human/Mouse/Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.