Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MAPK14 (pT180/Y182)/Pan MAPK14 (Human/Mouse) ELISA Kit

Catalog No. 89100450 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-450 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-450 Supplier Abnova™ Supplier No. KA2170

Includes

  • p38 alpha MAPK Microplate: 96 wells coated with anti-phospho-P38 alpha MAPK (Thr180/Tyr182) (half plate) and antip38 alpha MAPK antibody (half plate)
  • Wash Buffer Concentrate (20x)
  • Assay Diluent: 5x concentrated buffer
  • Detection Antibody p38 alpha MAPK: Rabbit anti-p38 alpha MAPK, 2 vial
  • HRP-conjugated Anti-rabbit IgG: 1,000x HRP-conjugated Anti-rabbit IgG concentrate, 25μL
  • TMB One-Step Substrate Reagent
  • Stop Solution: 0.2M sulfuric acid
  • Cell Lysate Buffer: 2x cell lysis buffer (not including protease and phosphatase inhibitors)
  • Positive Control HelaA003-1: Lyophilized powder from treated Hela cell lysate, 2 vials

Use to detect qualitatively human, mouse phospho MAPK14 (T180/Y182) and Pan MAPK14. Enzyme-linked immunosorbent assay.

MAPK14 (pT180/Y182)/Pan MAPK14 (Human/Mouse) ELISA Kit is a very sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates

Specifications

Conjugate HRP
Cross Reactivity Human, Mouse
Description MAPK14 (pT180/Y182)/Pan MAPK14 (Human/Mouse) ELISA Kit is an enzyme-linked immunosorbent assay for the qualitative determination of human, mouse phospho MAPK14 (T180/Y182) and Pan MAPK14.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Lysate
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Qualitative
Product Type MAPK14 (pT180/Y182)/Pan MAPK14 (Human/Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.