Learn More
Abnova™ MAPK3 (pT202/Y204)/MAPK1 (pT185/Y187) (Human) ELISA Kit
Shop All Abnova Corporation ProductsDescription
Includes
- Erk1/2Microplate: 96 wells coated with anti-pan Erk1/2 antibody
- Wash Buffer Concentrate (20x)
- Assay Diluent: 5x concentrated buffer. For diluting cell lysate sample, detection antibody and HRP-conjugated anti-rabbit IgG Concentrate
- Detection Antibody Erk1 (T202/Y204)/Erk2(T185/Y187): Rabbit anti-Erk1(T202/Y204)/ Erk2(T185/Y187)
- HRP-conjugated Anti-rabbit IgG: 500x HRP-conjugated anti-rabbit IgG, 25μL
- TMB One-Step Substrate Reagent
- Stop Solution: 0.2M sulfuric acid
- Cell Lysate Buffer: 2x cell lysis buffer (not including protease and phosphatase inhibitors)
- Positive Control A431S002-1: Lyophilized powder from A431 cell lysate
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Human |
| Description | MAPK3 (pT202/Y204)/MAPK1 (pT185/Y187) (Human) ELISA Kit is an enzyme-linked immunosorbent assay for the qualitative determination of human phospho MAPK3 (T202/Y204) and phospho MAPK1 (T185/Y187). |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Sample Type | Cell Lysate |
| Sample Volume | 100 μL |
| Packaging | 96-well |
| Content And Storage | Store the kit at -20°C |
| For Use With (Application) | Qualitative |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.