Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MB (Human) ELISA Kit

Catalog No. 89100015 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-015 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-015 Supplier Abnova™ Supplier No. KA0237
Add to Cart
Add to Cart

Includes

  • Antibody-Coated Wells; Microtiter well coated with mouse monoclonal anti-myoglobin, 96 wells
  • Reference Standard Set Contains 0, 25, 100, 250, 500, and 1000ng/mL myoglobin, liquid, ready-to-use. These standards have been pre-diluted 10-fold. Please do not dilute them again, 0.5mL each
  • Sample Diluent; Contains bovine serum and 1.0% (w/v) Pro-Clin as preservative, 25mL/bottle
  • Enzyme Conjugate Reagent, 22mL
  • TMB Reagent (One-Step), 11mL
  • Stop Solution (1N HCl), 11mL

Use to measure quantitatively myoglobin. Sandwich enzyme immunoassay.

The Myoglobin ELISA test is based on principle of a solid phase enzyme-linked immunosorbent assay

Specifications

Calibration Range 0 to 1000ng/mL
Conjugate HRP
Cross Reactivity Human
Description MB (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of myoglobin.
Detection Limit 5ng/mL
Detection Method Colorimetric
Includes Antibody-Coated Wells 96 wells, Reference Standard Set Contains 0, 25, 100, 250, 500, and 1000 ng/ml in. 0.5mL each, Sample Diluent (25mL), Enzyme Conjugate Reagent (22mL), TMB Reagent (One-Step) (11mL), Stop Solution (1N HCl) (11mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 20 μL
Sensitivity 5 ng/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.