Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mb (Mouse) ELISA Kit

Catalog No. 89100663 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Plasma, Serum
Plasma, Serum, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-663 Plasma, Serum, Urine
89-028-662 Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-663 Supplier Abnova™ Supplier No. KA2473
Add to Cart
Add to Cart

highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of MB in Mouse Serum or Plasma

Specifications

Conjugate HRP
Cross Reactivity Mouse
Description Mb (Mouse) ELISA Kit is for the quantitative measurement of Mb in plasma, serum and urine.
Detection Method Colorimetric
Includes Anti-Myoglobin-coated microtiter wells (96 wells), Mouse Myoglobin Stock (50μg/mL) (50μL), Diluent (12mL), Enzyme Conjugate Reagent (11mL), 20x Wash Solution (50mL), TMB Reagent (one step) (11mL), Stop Solution (1N HCl) (11mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum, Urine
Sample Volume 20 μL
Content And Storage Store lyophilized standard at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type Mb (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.