Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MB (Pig) ELISA Kit

Catalog No. 89100673 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-673 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-673 Supplier Abnova™ Supplier No. KA2483
Add to Cart
Add to Cart

Use to measure quantitatively plasma, serum and urine.

MB (Pig) ELISA Kit is an enzyme immunoassay for the quantitative determination of MB in pig serum, plasma and urine.

Quantification

Specifications

Conjugate HRP
Description MB (Pig) ELISA Kit is for the quantitative measurement of plasma, serum and urine.
Quantity 1 Kit
Content And Storage Store lyophilized standard at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type MB (Pig) ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.