Learn More
Description
MDA (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human MDA (methylene dioxyamphetamine)
Quantification
Specifications
Specifications
| Calibration Range | 0.312 to 20nM/mL |
| Conjugate | HRP |
| Detection Limit | 0.078nM/mL |
| Detection Method | Colorimetric |
| Includes | 1 Assay plate, standard 2, 20mL sample diluent, 10mL assay diluent A, 10mL assay diluent B, 120μL detection reagent A, 120μL detection reagent B, 30mL 25X Wash Buffer, 10mL Substrate, 10mL stop solution, 5 plate sealer |
| Quantity | 96 Assays |
| Sample Type | Biological Fluid, Plasma, Serum |
| Sample Volume | 50 μL |
| Content And Storage | 4°C |
| For Use With (Application) | Quantitative measurement of MDA (Methylene DioxyamphetAmine), quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.