Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Measles IgG ELISA Kit

Catalog No. 89100518 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-518 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-518 Supplier Abnova™ Supplier No. KA2256
Add to Cart
Add to Cart

Includes

  • Microwells coated with Measles antigen, 96 (12 x 8) wells
  • Sample Diluent: Ready to use, 22mL
  • Calibrator: Ready to use, 1mL
  • Positive Control: Ready to use, 1mL
  • Negative Control: Ready to use, 1mL
  • Enzyme conjugate: Ready to use, 12mL
  • TMB Substrate: Ready to use, 12mL
  • Stop Solution: Ready to use, 12mL
  • Wash concentrate 20X, 25mL

Detect IgG antibodies to Mumps in human serum or plasma.

Specifications

Conjugate HRP
Description Measles IgG ELISA Kit is intended for the detection of IgG antibodies to Mumps in human serum or plasma.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 10 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type Measles IgG ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.