Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Metanephrine ELISA Kit

Catalog No. 89028622 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-622 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-622 Supplier Abnova™ Supplier No. KA1889
Add to Cart
Add to Cart

Enzyme Immunoassay for the quantitative determination of free Metanephrine in Plasma

Specifications

Calibration Range 41 to 2100pg/mL
Conjugate IgG-Peroxidase
Description Metanephrine ELISA Kit ia an enzyme immunoassay for the quantitative determination of free Metanephrine in plasma.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 2 x 50 Reaction tubes, 4 adhesive foil, 20mL Wash Buffer concentrate, 12mL Enzyme Conjugate, 12mL Substrate, 12mL stop solution, 96-well Adrenaline-Metanephrine microtiter strip, 6mL Metanephrine Antiserum, Assay Buffer 12mL, 2 x 10mL equalizing reagent,
Quantity 1 Kit
Sample Type Plasma
Sample Volume 500 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Metanephrine ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.