Learn More
Specifications
Specifications
| Calibration Range | 40 to 1600ng/mL |
| Description | Metanephrine ELISA Kit ia an enzyme immunoassay for the quantitative determination of free Metanephrine in urine. |
| Detection Limit | 13ng/mL |
| Detection Method | Colorimetric |
| Includes | 2 x 50 Reaction tubes, Wash Buffer concentrate 1 x 20mL, Enzyme Conjugate 1 x 12mL, Substrate 1 x 12mL, stop solution 1 x 12mL, Adrenaline-Metanephrine microtiter strip 1 x 96-well, Metanephrine AntiSerum 1 x 12mL, Acylation concentrate 1 x 0.5mL, Acylation Diluent 1 x 4mL, Acylation Diluent 1 x 4mL, standard A 1 x 2mL,, standard B 1 x 2mL, standard C 1 x 2mL, standard D 1 x 2mL, standard E 1 x 2mL, standard F 1 x 2mL, Hydrochloric Acid 1 x 30mL,Control 1 1 x 2mL, Control 2 1 x 2mL, Acylation Buffer 1 x 30mL |
| Quantity | 1 Kit |
| Sample Type | Urine |
| Sample Volume | 25 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.