Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Metanephrine/Normetanephrine ELISA Kit

Catalog No. 89028611 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-611 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-611 Supplier Abnova™ Supplier No. KA1878
Add to Cart
Add to Cart

Enzyme immunoassay for the quantitative determination of free Metanephrine and free Normetanephrine in plasma

  • Sample Type: Plasma
  • Sample Volume: 500μL

Applications:

Quantification

Specifications

Calibration Range 41 to 2100pg/mL Metanephrine, 58 to 5800pg/mL Normetanephrine
Description Metanephrine/Normetanephrine ELISA Kit is an enzyme immunoassay for the quantitative determination of free Metanephrine and free Normetanephrine in plasma. This product contains two assay plates, for Metanephrine and Normetanephrine separately.
Detection Limit 17pg/mL Metanephrine, 23pg/mL Normetanephrine
Detection Method Colorimetric
Includes 2 x 50 Reaction tubes, Adhesive foil 2 x 4, Wash Buffer concentrate 2 x 20mL, Enzyme Conjugate 2 x 12mL,Substrate 2 x 12mL, stop solution 2 x 12mL, Adrenaline-Metanephrine microtiter strip 1 x 96-well, Noradrenaline-Normetanephrine microtiter strip 1 x 96-well, Metanephrine AntiSerum 1 x 6mL, Normetanephrine AntiSerum 1 x 6mL, Assay Buffer 1 x 12mL, Equalizing reagent 2 x 10mL, standard A 1 x 12mL, standard B 1 x 2mL, standard C 1 x 2mL, standard D 1 x 2mL, standard E 1 x 2mL, standard F 1 x 2mL, Acylation concentrate 1 x 1.5mL, Control 1 1 x 2mL, Control 2 1 x 2mL
Quantity 1 Kit
Sample Type Plasma
Sample Volume 500 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Metanephrine/Normetanephrine ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.