Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Metanephrine/Normetanephrine ELISA Kit

Catalog No. 89028612 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-612 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-612 Supplier Abnova™ Supplier No. KA1879
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enzyme Immunoassay for the quantitative determination of free Metanephrine and free Normetanephrine in Urine

Specifications

Calibration Range 40 to 1600pg/mL Metanephrine, 40 to 5200pg/mL Normetanephrine
Conjugate HRP
Description Metanephrine/Normetanephrine ELISA Kit is an enzyme immunoassay for the quantitative determination of free Metanephrine and free Normetanephrine in urine. This product contains two assay plates, for Metanephrine and Normetanephrine separately.
Detection Limit 5ng/mL Metanephrine, 20ng/mL Normetanephrine
Detection Method Colorimetric
Includes 2 x 50 Reaction tubes, Wash Buffer concentrate 1 x 20mL, Enzyme Conjugate 2 x 12mL, Substrate 2 x 12mL, stop solution 2 x 12mL, Adrenaline-Metanephrine microtiter strip 1 x 96-well, Noradrenaline-Normetanephrine microtiter strip 1 x 96-well, Normetanephrine AntiSerum 1 x 12mL, Metanephrine AntiSerum 1 x 12mL, Acylation concentrate 1 x 0.5mL, Acylation Diluent 1 x 4mL, standard A 1 x 2mL, standard B 1 x 2mL, standard C 1 x 2mL, standard D 1 x 2mL, standard E 1 x 2mL, standard E 1 x 2mL, Hydrochloric Acid 1 x 30mL, Control 1 1 x 2mL, Control 2 1 x 2mL, Acylation Buffer 1 x 30mL
Quantity 1 Kit
Sample Type Urine
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Metanephrine/Normetanephrine ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.