Learn More
Abnova™ Methyl-Histone H3-K9 ChIP Kit
Catalog No. 89027055 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-027-055 1 Kit
1 options
Catalog No. 89-027-055 Supplier Abnova™ Supplier No. KA0698
Add to Cart
Add to Cart
Methyl-Histone H3-K9 ChIP Kit is use for investigating the interactions of histone H3-K9 methylation and DNA in vivo efficiently.
  • The fastest procedure available, which can be finished within 5 hours
  • Strip microplate format makes the assay flexible: manual or high throughput
  • Columns for DNA purification are included: save time and reduce labor
  • Compatible with all DNA amplification-based approaches
  • Simple, reliable, and consistent assay conditions
  • Applications include combining the specificity of immunoprecipitation with qualitative and quantitative PCR, MS-PCR, DNA sequencing, and southern blot as well as DNA microarray
  • Store the following components at 4°C: Protease inhibitor cocktail, normal mouse IgG, Anti-dimethyl-H3-K9, proteinase K, control primers, 8-well assay strips; store other components at room temperature
  • The kit can be stable for up to 6 months from the shipment date, when stored properly
  • This kit has been configured for research use only and is not for diagnostic and clinical use
Specifications
Description Methyl-Histone H3-K9 ChIP Kit is use for investigating the interactions of histone H3-K9 methylation and DNA in vivo efficiently.
Content And Storage Store the following components at 4°C: Protease inhibitor cocktail, normal mouse IgG, Anti-dimethyl-H3-K9, proteinase K, 8 well strips Store other components at room temperature.
Quantity 1 Kit
For Use With (Application) Chromatin Immunoprecipitation
Type ChIP Kit