Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Methylated DNA Capture Kit

Catalog No. 89027048 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-048 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-048 Supplier Abnova™ Supplier No. KA0691
Add to Cart
Add to Cart

Used to enrich Methylated DNA

Specifications

Description Methylated DNA Capture Kit is used to enrich methylated DNA.
Includes 16mL MC1 (Antibody Buffer), 8mL MC2 (Reaction Buffer), MC3 (Wash Buffer) 2 x 16mL, 4mL MC4 (DNA release Buffer), 8mL MC5 (Binding Buffer), 1.2mL MC6 (Elution Buffer), 20μL Normal Mouse IgG (1mg/mL), 50μL Proteinase K (1
Quantity 1 Kit
Sample Type DNA
Content And Storage Store the following components at 4°C: MC1, MC3, normal mouse IgG, anti-5-methylcytosine, proteinase K, 8 well strips. Store other components at room temperature.
For Use With (Application) Methylated DNA Immunoprecipitation
Product Type Methylated DNA capture kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.