Learn More
Abnova™ Methylated DNA Immunoprecipitation Kit
Catalog No. 89027049 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-027-049 1 Kit
1 options
Catalog No. 89-027-049 Supplier Abnova™ Supplier No. KA0692
Add to Cart
Add to Cart
Methylated DNA Immunoprecipitation Kit is used to enrich methylated DNA from cell.
  • Quick Directly immunoprecipitate the methylated fractions of DNA from cell lysates
  • Highly efficient enrichment of methylated DNA: > 95%
  • The fastest procedure available, which can be finished within 4 hours
  • The Strip microplate format makes the assay flexible: manual or high throughput
  • Columns for DNA purification are included: save time and reduce labor
  • Compatible with all DNA amplification-based approaches
  • Simple, reliable, and consistent assay conditions
  • The Methylated DNA Immunoprecipitation Kit can be used for immunoprecipitating the methylated DNA from a broad range of species including human, rat, and mouse
  • The Methylated DNA Immunoprecipitation Kit is suitable for combining the specificity of methylated DNA immunoprecipitation with qualitative and quantitative PCR, and southern blot, as well as DNA microarray
  • Upon receipt store the following components at 4°C: normal mouse IgG, Proteinase K, Anti-5-Methylcytosine and 8-Well Assay Strip; store other components at room temperature
  • Kit can be stable for up to 6 months from the shipment date when stored properly
Specifications
Description Methylated DNA Immunoprecipitation Kit is used to enrich methylated DNA from cell.
Content And Storage Store the following components at 4°C: Normal mouse IgG, proteinase K, anti-5-methylcytosine, and 8 well strips Store other components at room temperature.
Quantity 1 Kit
For Use With (Application) Methylated DNA Immunoprecipitation
Type Immunoprecipitation/Co-Immunoprecipitation Kit