Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Methylated DNA Quantification Kit

Catalog No. 89027035 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-035 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-035 Supplier Abnova™ Supplier No. KA0676
Add to Cart
Add to Cart

Includes

  • SA1 (10X wash buffer), 30mL
  • SA2 (DNA binding solution), 3mL
  • SA3 (positive control, 100μg/mL), 20μL
  • SA4 (block solution), 20mL
  • SA5 (capture antibody, 1000μg/mL), 8μL
  • SA6 (labeling solution), 20μL
  • SA7 (fluoro developer), 24μL
  • SA8 (fluoro enhancer), 24μL
  • SA9 (fluoro dilution), 8mL
  • Negative control DNA (20ng/μL), 20μL
  • 8 well assay strips (with frame), 12

Used for measuring Methylated DNA using small amount of DNA samples

Specifications

Description Methylated DNA Quantification Kit is use for measuring methylated DNA using small amount of DNA samples.
Includes 30mL SA1 (Wash Buffer 10X), 3mL SA2 (DNA binding solution), 20μL SA3 (positive control, 100μg/mL), 20mL SA4 (block solution), 8μL SA5 (capture Antibody, 1000μg/mL), 20μ
Quantity 1 Kit
Sample Type DNA
Content And Storage Store SA3, SA6 SA7 and negative control DNA at -20°C away from light. Store SA1, SA4, SA5, SA8 and 8 well strips at 4°C away from light. Store other components at room temperature.
For Use With (Application) Quantification
Product Type Methylated DNA Quantification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.