Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MICA (Human) ELISA Kit

Catalog No. 89100790 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma, Serum, Tissue
Cell Lysates, Tissue Lysates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-790 Cell Lysates, Tissue Lysates
89-027-121 CCS, Plasma, Serum, Tissue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-790 Supplier Abnova™ Supplier No. KA3087
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human MICA

Specifications

Calibration Range 62.5 to 4000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description MICA (Human) ELISA Kit is intended for the quantitative measurement of human MICA.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Human MICA antibody, Lyophilized MICA standards (10ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Hman MICA antibody (Concentrated Dilution: 1:100) (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (ConcentratedDilution: 1:100) (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Cell Lysates, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type MICA (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.