Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Microcystin-LR ELISA Kit

Catalog No. 89028336 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-336 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-336 Supplier Abnova™ Supplier No. KA1496
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • ELISA strips (colourless) coated with specific antibody, 96-wells
  • Standard A, Ready to use, 0μg/L, 0.6mL
  • Standard B, Ready to use, 0.1μg/L, 0.6mL
  • Standard C, Ready to use, 0.5μg/L, 0.6mL
  • Standard D, Ready to use, 1μg/L, 0.6mL
  • Standard E, Ready to use, 2.5μg/L, 0.6mL
  • MC-LR labeled with horseradish peroxidase, Ready to use, (Px-conjugate), Ready to use, 2.5mL
  • Wash buffer concentrate, 10x concentrated, 125mL
  • Dilution buffer, Ready to use, 15mL
  • Chromogenic substrate (TMB substrate), Ready to use, 13mL
  • Stop solution, Ready to use, 15mL
  • Sealable pouch for unused strips, 1 piece

Intended for the detection of Microcystin - LR (MC-LR) in environmental samples-potable water and surface water samples.

Specifications

Conjugate HRP
Description Microcystin-LR ELISA Kit is a competitive binding enzyme immunoassay for the quantitative measurement of Microcystin-LR.
Detection Limit 0.1μg/L
Detection Method Colorimetric
Includes 8-well break away strip coated with the Antibody 96-well, 0.6mL MC-LR standard A-E, 2.5mL MC-LR-Px Enzyme Conjugate, 125mL Wash Buffer (10X), 125mL Dilution Buffer, 15mL Chromogenic Substrate, 30mL stop solution
Quantity 1 Kit
Sample Type Water
Sample Volume 100 μL
Content And Storage Store at 4°C on dry atmosphere away from light.
For Use With (Application) Quantification
Product Type Microcystin-LR ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.