Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MMP13 (Human) ELISA Kit

Catalog No. 89100794 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-794 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-794 Supplier Abnova™ Supplier No. KA3091
Add to Cart
Add to Cart

Highly sensitive and specific quantitative determination of activated Human MMP13

Specifications

Calibration Range 78 to 5000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description MMP13 (Human) ELISA Kit is intended for the quantitative measurement of human MMP13.
Detection Limit 5pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Human MMP13 antibody, Lyophilized MMP13 standards (10ng x 2), Standard diluent buffer (30mL), Biotin conjugated anti-Human MMP13 antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (130uL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), 25X Wash buffer (30mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type MMP13 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.