Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MMP7 (Human) ELISA Kit

Catalog No. 89100288 Shop All Abnova Corporation Products
Change view
Click to view available options
Conjugate:
Biotin
HRP
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
89-100-288 Biotin
89-100-792 HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-288 Supplier Abnova™ Supplier No. KA0395
Add to Cart
Add to Cart

Includes

  • Lyophilized recombinant human MMP7 standard: 10ng/tube x 2
  • One 96-well plate precoated with anti-human MMP7 antibody
  • Sample diluent buffer: 30mL
  • Biotinylated anti-human MMP7 antibody: 130μL, dilution 1:100
  • Antibody diluent buffer: 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC): 130μL, dilution 1:100
  • ABC diluent buffer: 12mL
  • TMB color developing agent: 10mL
  • TMB stop solution: 10mL

The ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description MMP7 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human MMP7.
Detection Limit One 96-well plate
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Lyophilized recombinant Human MMP7 standard (10ng x 2 vials), Sample diluent buffer (30mL), Biotinylated anti-Human MMP7 antibody (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase complex (ABC): 130μL, ABC diluent buffer (12mL), TMB color developing agent (10mL), TMB stop solution (10mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Sensitivity <6 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type ELISA Kit
Specificity No detectable cross-reactivity with any other cytokine
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.