Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MPO (Human) ELISA Kit

Catalog No. 89100795 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Serum, Tissue Lysates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-795 Body Fluid, CCS, Serum, Tissue Lysates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-795 Supplier Abnova™ Supplier No. KA3092
Only null left
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-Human Myeloperoxidase/MPO antibody, 96 (8x12) wells
  • Lyophilized Myeloperoxidase / MPO standards, 10ng x 2
  • Sample / Standard diluent buffer, 30mL
  • Biotin conjugated anti-Human Myeloperoxidase / MPO antibody (Concentrated), Dilution: 1:100, 130μL
  • Antibody diluent buffer, 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
  • ABC diluent buffer, 12mL
  • TMB substrate, 10mL
  • Stop solution, 10mL
  • Wash buffer (25X), 30mL

Use to measure quantitatively human MPO.

This kit is based on sandwich enzyme-linked immune-sorbent assay technology.

Specifications

Calibration Range 312 to 20000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description MPO (Human) ELISA Kit is intended for the quantitative measurement of human MPO.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-Human Myeloperoxidase / MPO antibody 96 (8x12) wells, Lyophilized Myeloperoxidase / MPO standards (10ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Human Myeloperoxidase / MPO antibody (Concentrated), Dilution: 1:100 (130mL), Antibody diluent buffer: 12mL, Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Serum, Tissue Lysates
Sample Volume 100 μL
Sensitivity <10 pg/mL
Packaging 96-well
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type MPO (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.