Learn More
Description
Includes
- Microwell coated with Mumps antigen, 12x8x1
- Sample Diluent: 1 bottle (ready to use), 22mL
- Calibrator: 1 Vial (ready to use), 1.5mL
- Positive Control: 1 vial (ready to use), 1.5mL
- Negative Control: 1 vial (ready to use), 1.5mL
- Enzyme conjugate: 1 bottle (ready to use), 12mL
- TMB Substrate: 1 bottle (ready to use), 12mL
- Stop Solution: 1 bottle (ready to use), 12mL
- Wash concentrate 20X: 1 bottle, 25mL
Specifications
Specifications
| Conjugate | HRP |
| Description | Mumps IgM ELISA Kit is intended for the detection of IgG class antibodies to Measles in human serum or plasma |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Sample Type | Plasma, Serum |
| Sample Volume | 10 μL |
| Packaging | 96-well |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Qualitative |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.