Learn More
Specifications
Specifications
| Conjugate | HRP |
| Description | Mycoplasma pneumoniae IgA ELISA Kit is a solid phase enzyme immunoassay for the semiquantitative measurement of IgA autoantibodies against Mycoplasma pneumoniae. |
| Detection Method | Colorimetric |
| Includes | 96-well ELISA break away strip coated with specific Antigen, 1.3mL negative Control Serum, 1.3mL positive Control Serum, 1.3mL calibrator, 0.2mL Antibodies-Px Conjugate,125mL Wash Buffer concentrate, 125mL Dilution Buffer, 15mL Chromogenic Substrate, 15mL stop solution |
| Quantity | 1 Kit |
| Sample Type | Serum |
| Sample Volume | 100 μL |
| Content And Storage | Store at 4°C on dry atmosphere away from light. |
| For Use With (Application) | Quantification |
| Product Type | Mycoplasma pneumoniae IgA ELISA Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.