Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mycoplasma pneumoniae IgM ELISA Kit

Catalog No. 89100523 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-523 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-523 Supplier Abnova™ Supplier No. KA2261
Add to Cart
Add to Cart

Detection of IgM antibodies to mycoplasma pneumoniae in Human serum or plasma

Specifications

Conjugate HRP
Cross Reactivity Human
Description Mycoplasma pneumoniae IgG ELISA Kit isintended for the detection of IgM antibodies to Mycoplasma pneumoniae in human serum or plasma.
Detection Method Colorimetric
Includes 96-well Microwell strips, Sample Diluent (22mL), Calibrator (1.5mL), Positive control (1.5mL), Negative control (1.5mL), Enzyme conjugate (12mL), TMB color developing agent (12mL), Stop Solution (12mL), 20X wash conc. (25mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type Mycoplasma pneumoniae IgM ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.