Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ NFKB Phosphorylation Cell-Based ELISA Kit

Catalog No. 89101091 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-091 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-091 Supplier Abnova™ Supplier No. KA3792
Add to Cart
Add to Cart

Includes

  • 10X Wash Buffer, 25mL
  • Protein Stain, 6mL
  • pNFkB-Ab1, 10μL
  • Blocking Buffer, 25mL
  • HRP Substrate, 6mL
  • HRP-Ab2, 10μL

Use to detect qualitatively NFKB phosphorylation in cultured cells. Indirect enzyme-linked immunoassay.

  • New and Improved. Total assay time reduced from typical 9 hours to 5 hours
  • Simple. No cell lysis necessary. Cells can be cultured in 96-well plates
  • Convenient. Total protein and pNFkB can be measured in the same sample

Specifications

Cross Reactivity Human, Mouse
Description NFKB Phosphorylation Assay Kit is an indirect enzyme-linked immunoassay for qualitative determination of NFKB phosphorylation in cultured cells.
Detection Method Fluorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell
Total Assay Time 5 hr.
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 100
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.