Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Norepinephrine ELISA Kit

Catalog No. 89028624 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Biological Fluid
Plasma, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-028-624 Plasma, Urine
89-101-135 Biological Fluid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-624 Supplier Abnova™ Supplier No. KA1891
Add to Cart
Add to Cart

Includes

  • Adhesive foil ready for use, 1 x 4
  • Wash buffer concentrate, Concentrate. Dilute content with dist. Water to a final volume of 1000mL, 1 x 20mL
  • Enzyme conjugate ready for use, anti-rabbit IgG conjugated with peroxidase, 1 x 12mL
  • Substrate ready for use, containing a solution of tetramethylbenzidine (TMB), 1 x 12mL
  • Stop solution ready for use, containing 0.25M H2SO4, 1 x 12mL
  • Noradrenaline-Normetanephrine Microtiter Strips 12 strips, 8 wells each, break apart, pre-coated, yellow coloured, 1 x 96 wells
  • Noradrenaline antiserum from rabbit, ready for use, yellow coloured, yellow screw cap, 1 x 6mL
  • Adjustment buffer ready for use, 1 x 4mL
  • Standard A ready for use, 1 x 4mL
  • Standard B ready for use, 1 x 4mL
  • Standard C ready for use, 1 x 4mL
  • Standard D ready for use, 1 x 4mL
  • Standard E ready for use, 1 x 4mL

Enzyme Immunoassay for the quantitative determination of Norepinephrine in Plasma and Urine

Specifications

Calibration Range 20 to 339ng/mL Urine, 318 to 2436pg/mL Plasma
Description Norepinephrine ELISA Kit is an enzyme immunoassay for the quantitative determination of Norepinephrine in plasma and urine.
Detection Limit 1.5ng/mL Urine, 50pg/mL Plasma
Detection Method Colorimetric
Includes Adhesive foil 1 x 4, Wash Buffer concentrate 1 x 20mL, Enzyme Conjugate 1 x 12mL, Substrate 1 x 12mL, stop solution 1 x 12mL, Noradrenaline-Normetanephrine microtiter strip 1 x 96-well, Noradrenaline antiserum 1 x 6mL, Adjustment Buffer 1 x 4mL, standard
Quantity 1 Kit
Sample Type Plasma, Urine
Sample Volume Urine: 10 μL; Plasma: 300 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Norepinephrine ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.