Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Normetanephrine ELISA Kit

Catalog No. 89028625 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-625 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-625 Supplier Abnova™ Supplier No. KA1892
Add to Cart
Add to Cart

Enzyme Immunoassay for the quantitative determination of free Normetanephrine in Plasma

Specifications

Calibration Range 58 to 5800pg/mL
Description Normetanephrine ELISA Kit is an enzyme immunoassay for the quantitative determination of free Normetanephrine in plasma.
Detection Limit 23pg/mL
Detection Method Colorimetric
Includes 2 x 50 Reaction tubes, Adhesive foil 1 x 4, Wash Buffer concentrate 1 x 20mL, Enzyme Conjugate 1 x 12mL, Substrate 1 x 12mL, stop solution 1 x 12mL, Normetanephrine-Normetanephrine microtiter strip 1 x 96-well, Normetanephrine antiserum 1 x 6mL, Assay Buffer 1 x 12mL, Equalizing reagent 2 x 10mL, standard A 1 x 12mL, standard B 1 x 2mL,standard C 1 x 2mL, standard D 1 x 2mL, standard E 1 x 2mL, standard F 1 x 2mL, Acylation concentrate 1 x 1.5mL, Control 1 1 x 2mL, Control 2 1 x 2mL
Quantity 1 Kit
Sample Type Plasma
Sample Volume 500 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Normetanephrine ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.