Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ NPPA (Human/Mouse/Rat) ELISA Kit

Catalog No. 89028416 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-416 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-416 Supplier Abnova™ Supplier No. KA1680

Includes

  • ANP Microplate (Item A): coated with secondary antibody. 96 wells (12 strips x 8 wells)
  • Wash Buffer Concentrate (20x) (Item B), 25mL
  • Lyophilized Standard ANP Peptide (Item C), 2 vials
  • Lyophilized anti-ANP polyclonal antibody (Item N), 2 vials
  • 1x Assay Diluent E (Item R): Diluent for both standards and samples including serum or plasma, cell culture media or other sample types, 25mL x 2
  • Lyophilized Biotinylated ANP peptide, (Item F), 2 vials
  • HRP-Streptavidin concentrate (Item G): 50x concentrated HRP-conjugated Streptavidin, 600μL
  • Lyophilized Positive control (Item M), 1 vial
  • TMB One-Step Substrate Reagent (Item H): 3, 3', 5, 5'-tetramethylbenzidine (TMB) in buffered solution, 12mL
  • Stop Solution (Item I): 0.2M sulfuric acid, 8mL

For detecting NPPA/Nppa based on the principle of competitive Enzyme Immunoassay

Specifications

Calibration Range 0.1 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Human, Mouse, Rat
Description NPPA/Nppa (Human, Mouse, Rat) ELISA Kit is an in vitro quantitative assay for detecting NPPA/Nppa based on the principle of Competitive Enzyme Immunoassay.
Detection Limit 2.3pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 25mL 20X) Wash Buffer concentrated, detected Polyclonal Antibody (2 vials), standard (2 vials), 30mL assay diluent A, 15mL assay diluent B, Biotinylated peptide (2 vials), 8μL HRP-Streptavidian concentr
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Serum, Synovial Fluid
Sample Volume 100 μL
Content And Storage Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type NPPA (Human/Mouse/Rat) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.