Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Nppb (Rat) ELISA Kit

Catalog No. p-4392261 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
0.031 to 1ng/mL
0.031 to 2ng/mL
Quantity:
1 Kit
96 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range Quantity
89-028-040 0.031 to 1ng/mL 96 Assays
89-100-220 0.031 to 2ng/mL 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-040 Supplier Abnova™ Supplier No. KA0979
Only null left
Add to Cart
Add to Cart

Includes

  • rBNP-45 Microplate: A 96 well polystyrene microplate (12 strips of 8 wells) coated with a polyclonal antibody against rBNP-45, 1 in no.
  • Sealing Tapes: pressure-sensitive sealing tapes, which can be cut to fit the format of the individual assay, 3 in nos.
  • rBNP45 Standard: Rat BNP-45 in a buffered protein base (lyophilized), 4ng
  • Biotinylated rBNP-45 Antibody (100x): A 100-fold biotinylated polyclonal antibody against rBNP-45, 80μL
  • MIX Diluent Concentrate (10x): A 10-fold buffered protein base, 30mL
  • Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant, 30mL, 2 bottles
  • Streptavidin-Peroxidase Conjugate: A 100-fold concentrate 80μL
  • Ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine, 8mL
  • Stop Solution: A 0.5N hydroxychloric acid to stop the chromogen substrate reaction, 12mL

For detection of Rat Nppb in Rat Plasma, Serum, tissue extract and cell culture supernatants

Specifications

Calibration Range 0.031 to 1ng/mL
Conjugate Biotin
Description Nppb (Rat) ELISA Kit is designed for detection of rat Nppb in rat plasma, serum, tissue extract, and cell culture supernatants.
Detection Limit 0.03ng/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 3ng rBNP45 standard, 100X Biotinylated rBNP45 Antibody 1 vial, 30mL MIX diluent concentrate (10X), Wash Buffer 20X 30mL x 2, Streptavidin-peroxidase Conjugate 80, 8mL Chromogen Substrate, 12mL stop solution
Quantity 96 Assays
Sample Type CCS, Plasma, Serum, Tissue
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Nppb (Rat) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.