Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ NTF3 (Human) ELISA Kit

Catalog No. 89022844 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-844 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-844 Supplier Abnova™ Supplier No. KA0402
Only null left
Add to Cart
Add to Cart

Includes

  • Lyophilized recombinant human NT-3 standard 10ng/tube+2
  • One 96-well plate precoated with anti-human NT-3 antibody 1
  • Sample diluent buffer 30mL
  • Biotinylated anti-human NT-3 antibody, dilution 1:100 130μL
  • Antibody diluent buffer 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100 130μL
  • ABC diluent buffer 12mL
  • TMB color developing agent 10mL
  • TMB stop solution 10mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Human NTF3

Specifications

Calibration Range 15.6 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description NTF3 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human NTF3.
Detection Limit 2pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, lyophilized recombinant Human NTF3 standard 10ng x 2 vials, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human NTF3 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin-Biotin-P
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type NTF3 (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.