Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ NTF4 (Rat) ELISA Kit

Catalog No. 89100800 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-800 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-800 Supplier Abnova™ Supplier No. KA3097
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-rat NT-4 antibody, 96 (8 x 12) wells
  • Standard (22500pg/mL), 0.5mL
  • Standard diluent buffer, 1.5mL
  • Wash buffer (30x), Dilution: 1:30, 20mL
  • Sample diluent buffer, 6mL
  • HRP conjugated anti-rat NT-4 antibody (RTU), 6mL
  • Stop solution, 6mL
  • TMB substrate A, 6mL
  • TMB substrate B, 6mL
  • Plate sealer, 2 slices
  • Hermetic bag, 1 bag

Use to measure quantitatively rat Ntf4.

This kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 600 to 20,000pg/mL
Conjugate HRP
Cross Reactivity Rat
Description Ntf4 (Rat) ELISA Kit is intended for the quantitative measurement of rat Ntf4.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Sample, Urine
Sample Volume 10 μL
Packaging 96-well
Content And Storage Store at 4°C for 6 months.
For Use With (Application) Quantification
Product Type Ntf4 (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.