Learn More
Abnova™ One-Step ChIP Kit
Catalog No. 89028209 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-028-209 1 Kit
1 options
Catalog No. 89-028-209 Supplier Abnova™ Supplier No. KA1514
Add to Cart
Add to Cart
One-Step ChIP Kit is use for investigating interactions of proteins and DNA.
  • The fastest and most convenient ChIP method: entire procedure (from intact chromatin sample to ready-for-use DNA) is less than 60 minutes with the actual handling time being less than 10 minutes due to simultaneous processing of chromatin shearing and immunoprecipitation (One-Step ChIP)
  • 96-well plate format makes the assay flexible; either (a) manual with one single reaction each time; or (b) high throughput with 96 reactions each time.
  • Highly efficient enrichment: enrichment ratio of positive to negative control > 120, and an extremely low number of cells required (as low as 10,000 cells per ChIP reaction)
  • Wide downstream analysis compatibility: compatible with various downstream analysis workflows including ChIP-PCR, ChIP-on-chip, and ChIP-seq
  • Suitable for selective enrichment of a chromatin fraction containing specific DNA sequences in a high throughput format using chromatin solated from various species, particularly mammals
  • Kit has been configured for research use only
  • Kit is shipped in two parts: the first at ambient room temperature and the second part on frozen ice packs at 4°C
  • All reagents are stable for 6 months under proper storage conditions
Specifications
Description One-Step ChIP Kit is use for investigating interactions of proteins and DNA.
Content And Storage Store CH1, Non-Immune IgG, Anti-RNA Polymerase II, Proteinase K, GAPDH Primer-Forward, GAPDH Primer-Reverse, and 8-Well Assay Strips (With 1 Frame) at 4°C away from light. Store all other components at room temperature away from light.
Quantity 1 Kit
For Use With (Application) Chromatin Immunoprecipitation
Type ChIP Kit