Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ORANGE Peroxidase Substrate Kit

Catalog No. 89027127 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-127 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-127 Supplier Abnova™ Supplier No. KA0563
Add to Cart
Add to Cart

Immunohistochemistry

Specifications

Description Metal enhanced DAB technology permits detection of extremely low concetrations of tissue antigens. This kit provides a sensitive means for detecting peroxidase labeled antibodies and conjugates. ORANGE Peroxidase Substrate Kit contains nickel enhanced DAB that produces an orange-red DAB polymer and also exceeds the sensitivity of DAB alone.
Includes 50mL Enhance ORANGE Buffer solution, 10mL DAB-C solution, 10mL Peroxide solution, 50mL Contrast GREEN solution, 10mL blocking solution concentrate (10X)
Quantity 1 Kit
Sample Type Tissue
Content And Storage Store the kit at 4°C
For Use With (Application) Immunohistochemistry
Product Type Orange Peroxidase Substrate kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.