Learn More
Specifications
Specifications
| Conjugate | HRP |
| Description | Ochratoxin-A ELISA Kit is a competitive binding enzyme immunoassay for the quantitative measurement of ochratoxin-A. |
| Detection Method | Colorimetric |
| Includes | 96-well microtiter plate, 0.9mL negative calibrator, positive calibrator 3 vials x 0.75mL, 10.5mL Ochratoxin-A-HRP Conjugate (ORT-HRP), 10.5mL Etramethylbenzidine (TMB) Substrate, 15mL Wash Buffer, 10.5mL stop solution |
| Quantity | 1 Kit |
| Sample Type | Animal Feed, Cereal, Coffee, Grain |
| Sample Volume | 25 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Product Type | Ochratoxin-A ELISA Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.