Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ORM1 (Rat) ELISA Kit

Catalog No. 89100680 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Biological Fluid
Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-680 Serum
89-100-402 Biological Fluid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-680 Supplier Abnova™ Supplier No. KA2490
Add to Cart
Add to Cart

Includes

  • Anti-rat α-1-AGP-coated wells, 96 wells (8 x 12 strips)
  • Rat α-1-AGP stock, 2μg/mL (Lyophilized)
  • 10X Diluent, 25mL
  • Enzyme Conjugate Reagent, 11mL
  • 20X Wash Solution, 50mL
  • TMB Reagent (One step), 11mL
  • Stop Solution (1N HCl), 11mL

Orm1 (Rat) ELISA Kit is based on a solid phase enzyme-linked immunosorbent assay (ELISA)

Specifications

Conjugate HRP
Cross Reactivity Rat
Description Orm1 (Rat) ELISA Kit is for the quantitative measurement of Orm1 in serum.
Detection Limit 1.56 to 100ng/mL
Detection Method Colorimetric
Includes Anti-rat α -1-AGP-coated wells (96 wells (8 x 12 strips), Rat α-1-AGP stock (2μg/mL (Lyophilized), 10 x diluent (25mL), Enzyme conjugate reagent (11mL), 20 x wash solution (50mL), TMB reagent (one step): 11mL, Stop solution (1N HCl): 11mL
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Orm1 (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.