Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PCP ELISA Kit

Catalog No. 89024479 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-479 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-479 Supplier Abnova™ Supplier No. KA0938
Add to Cart
Add to Cart

Specific and sensitive in-vitro test to detect the presence of PCP in samples such as whole Blood, oral fluids, Serum, Plasma and Urine

Specifications

Calibration Range 2 to 10ng/mL
Conjugate HRP
Description PCP ELISA Kit is a specific and sensitive in-vitro test to detect the presence of PCP in samples such as whole blood, oral fluids, serum, plasma and urine.
Detection Limit 0.5ng/mL
Detection Method Colorimetric
Includes Microwell strip 12 x 8 x 1, 12.5mL Enzyme Conjugate, 1mL positive reference standard, 1mL negative control, 14mL TMB chromogenic Substrate, 12.5mL stop solution
Quantity 96 Tests
Sample Type Oral Fluid, Plasma, Serum, Urine, Whole Blood
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type PCP ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.