Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PDGFRB (Human) ELISA Kit

Catalog No. 89100422 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-422 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-422 Supplier Abnova™ Supplier No. KA2142

Includes

  • PDGFR-beta Microplate (Item A): coated with anti-human PDGFR-beta antibody
  • Wash Buffer Concentrate (20x) (Item B)
  • Standard Protein (Item C): Human PDGFR-beta is enough to rum each standard in duplicate
  • Assay Diluent D (Item K): 5x concentrated buffer
  • Assay Diluent B (Item E): 5x concentrated buffer
  • Detection Antibody PDGFR-beta (Item F): Biotinylated anti-human PDGFR-beta, 2 vial
  • HRP-Streptavidin Concentrate (Item G): 300x concentrated HRP-conjugated streptavidin
  • TMB One-Step Substrate Reagent (Item H): 3,3',5,5'-tetramethylbenzidine (TMB) in buffered solution
  • Stop Solution (Item I): 0.2M sulfuric acid

Use to measure quantitatively human PDGFRB. Enzyme-linked immunosorbent assay.

This assay employs an antibody specific for human PDGFR-beta coated on a 96-well plate

Specifications

Conjugate HRP
Cross Reactivity Human
Description PDGFRB (Human) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of human PDGFRB.
Detection Limit 25pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Serum, Plasma, Cell Culture Supernatant
Sample Volume 100 μL
Sensitivity 25 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type PDGFRB (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.