Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PGFM ELISA Kit

Catalog No. 89100928 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-928 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-928 Supplier Abnova™ Supplier No. KA3391
Add to Cart
Add to Cart

Includes

  • Coated Clear 96 Well Plate: A clear plastic microtiter plate with break-apart strips, coated with goat anti-rabbit IgG
  • PGFM Standard: 13,14-dihydro-15-keto-PGF2ɑ at 32,000pg/mL in a special stabilizing solution
  • PGFM Antibody: A rabbit polyclonal antibody specific for PGFM, 3mL
  • PGFM Conjugate: A PGFM-peroxidase conjugate in a special stabilizing solution, 3mL
  • Assay Buffer Concentrate: A 5X concentrate that must be diluted with deionized or distilled water, 28mL
  • Wash Buffer Concentrate: A 20X concentrate
  • TMB Substrate
  • Stop Solution: A 1M solution of hydrochloric acid. CAUSTIC, 5mL
  • Plate Sealer, 1 slice

Use to detect quantitatively of 13,14-dihydro-15-keto-PGF-2 alpha (PGFM) in fecal extracts, urine, serum and plasma samples. Enzyme-linked immunosorbent assay.

Specifications

Calibration Range 50 to 3200pg/mL
Conjugate HRP
Description PGFM ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of 13,14-dihydro-15-keto-PGF-2 alpha (PGFM) in fecal extracts, urine, serum and plasma samples.
Detection Limit 46.2pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Fecal Extract, Plasma, Serum, Tissue Culture Medium, Urine
Sample Volume 50 μL
Sensitivity 20.8 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type PGFM ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.