Learn More
Specifications
Specifications
| Calibration Range | 6.3 to 100μ/mL |
| Conjugate | HRP |
| Description | Phosphatidyl Inositol Ab IgG/IgM ELISA Kit is an indirect solid phase ELISA for the quantitative measurement of IgG and IgM class autoantibodies against phosphatidyl inositol in human serum or plasma. |
| Detection Method | Colorimetric |
| Includes | 1.5mL Combined calibrator with IgG and IgM class Anti-Phospholipid Antibodies 6 vials, 1.5mL Anti-Phospholipid Controls 2 vials, 20mL Sample Buffer 1 vial, 15mL Enzyme Conjugate solution 1 vial, 15mL TMB Substrate solution 1 vial, 15mL stop solution 1 vial, 96-well plate 1, 20mL Wash solution 1 vial |
| Quantity | 96 Tests |
| Sample Type | Plasma, Serum |
| Sample Volume | 10 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.