Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Phospholipid Screen IgG/IgM ELISA Kit

Catalog No. 89027247 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-247 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-247 Supplier Abnova™ Supplier No. KA1109
Add to Cart
Add to Cart

For IgG and IgM autoantibodies against Cardiolipin, Phosphatidyl Ser, Phosphatidyl Inositol and Phosphatidic Acid in Human Serum, Plasma

Specifications

Calibration Range 6.3 to 100U/mL
Conjugate HRP
Description Phospholipid Screen IgG/IgM ELISA Kit is a quantitative assay intended to screen for the presence of IgG and IgM class autoantibodies against Cardiolipin, Phosphatidyl Serine, Phosphatidyl Inositol and Phosphatidic acid in human serum or plasma.
Detection Limit 0.5U/mL
Detection Method Colorimetric
Includes 1.5mL Combined Anti-phospholipid calibrator in a Serum/Buffer Matrix 6 vial, 1.5mL Anti-phospholipid controls in a Serum/Buffer Matrix 2 vial, 20mL Sample Buffer 1 vial, 15mL Enzyme Conjugate solution 1 vial, 15mL TMB Substrate solution 1 vial, 15mL stop
Quantity 96 Assays
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Phospholipid Screen IgG/IgM ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.