Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Phosphorylated NEFH (Human) ELISA Kit

Catalog No. 89028319 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-319 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-319 Supplier Abnova™ Supplier No. KA1479
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Phosphorylated NEFH

Specifications

Conjugate HRP
Description Phosphorylated NEFH (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of phosphorylated NEFH.
Detection Limit 24pg/mL
Detection Method Colorimetric
Includes Microtitre plate with immobilised Antibody NF01 (96-well), 0.1mL pNF-H standard, 0.1mL NF05-HRP (100X), Detection Antibody Conjugated to HRP (100X), 125mL Wash Buffer (10X), 15mL Dilution Buffer, 25mL Chromogenic Substrate, 15mL stop solution
Quantity 1 Kit
Sample Type Cerebrospinal Fluid, Plasma, Serum
Sample Volume 25 μL
Content And Storage Store at 4°C on dry atmosphere away from light.
For Use With (Application) Quantification
Product Type Phosphorylated NEFH (Human) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.