Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Plant Total RNA Purification Kit

Catalog No. 89028632 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-632 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-632 Supplier Abnova™ Supplier No. U0065
Add to Cart
Add to Cart

Plant Total RNA Purification Kit is designed specifically for purifying total RNA from plant tissue and cells.

  • Provides an efficient method for purifying total RNA from plant tissue and cells
  • Samples are ground in liquid nitrogen and then filtered to remove cell debris
  • In the presence of a binding buffer and chaotropic salt, total RNA in the lysate binds to the glass fiber matrix of the spin column
  • Optional DNase treatments can be followed to remove unwanted DNA residue
  • The procedure does not require phenol extraction or alcohol precipitation and can be completed within 30 minutes
  • The purified total RNA is ready for use in RT, RT-PCR, Real-time PCR and Northern Blotting
  • Store at room temperature

Specifications

Content And Storage Store at room temperature.
Description Plant Total RNA Purification Kit is designed specifically for purifying total RNA from plant tissue and cells.
For Use With (Application) Purification
Includes 60mL RB Buffer, 60mL PRB Buffer, 50mL W1 Buffer, 25mL Wash Buffer (add 100mL Ethanol), 15mL RNase-free Water, filter column, RB column, 2mL collection tube
Product Type Plant Total RNA Purification Kit
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.