Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PLAT (Human), Biotin, ELISA Kit

Catalog No. 89101130 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-130 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-130 Supplier Abnova™ Supplier No. KA3834
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Human tPA Microplate: A 96 well polystyrene microplate coated with a polyclonal antibody against tPA. 96 wells (12 strips of 8 wells)
  • Sealing Tapes: Each kit contains 3 precut, pressure sensitive sealing tapes that can be cut to fit the format of the individual assay. 3 Human tPA Standard: Human tPA in a buffered protein base (lyophilized). 10ng
  • Biotinylated Human tPA antibody (50x): A 50-fold concentrated biotinylated antibody against human tPA. 140μL
  • MIX Diluent Concentrate (10x): A 10-fold concentrated buffered protein base. 30mL
  • Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant. 30mL x 2
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate. 80μL
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine. 8mL
  • Stop Solution: A 0.5 N hydrochloric acid to stop the chromogen substrate reaction. 12mL

PLAT (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human PLAT.

  • Kit is designed for detection of human tPA in plasma, serum, milk, cell culture supernatants, and tissue extract
  • This assay employs a quantitative sandwich enzyme immunoassay technique that measures total tPA in less than 4 hours
  • Store components of the kit at 2-8°C or -20°C upon arrival to the expiration date
  • For research use only

Specifications

Conjugate Biotin
Cross Reactivity Human, Pig
Description PLAT (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human PLAT.
Detection Limit 0.1 ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell culture supernatant, milk, plasma, serum and tissue extract
Sample Volume 50 μL
Content And Storage Store the kit at 4°C. For long term storage, please store at -20°C.
Form Lyophilized
For Use With (Application) Quantification
Product Type PLAT (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.