Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Plg (Rat) ELISA Kit

Catalog No. 89101127 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Biological Sample
CCS, Plasma, Serum, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-101-127 CCS, Plasma, Serum, Urine
89-028-675 Biological Sample
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-127 Supplier Abnova™ Supplier No. KA3831
Add to Cart
Add to Cart

highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of PLG in Rat biological fluids

Specifications

Conjugate Biotin
Cross Reactivity Rat
Description Plg (Rat) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of rat Plg.
Detection Limit 0.3ng/ml
Detection Method Colorimetric
Includes 96 wells Rat Plasminogen Microplate, 3 Sealing Tapes, Rat Plasminogen Standard (800ng), 70X Biotinylated Rat Plasminogen Antibody (105uL), 10X EIA Diluent Concentrate (30mL), 20X Wash Buffer Concentrate (2 x 30mL), Streptavidin-Peroxidase Conjugate (SP Conjugate) (80uL), Chromogen Substrate (8mL), Stop Solution (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum, Urine
Sample Volume 50 μL
Content And Storage Store the kit at 4°C. For long term storage, please store at -20°C.
For Use With (Application) Quantification
Product Type PLG (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.