Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PNPLA3 (Human/Mouse/Rat) ELISA Kit

Catalog No. 89028414 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-414 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-414 Supplier Abnova™ Supplier No. KA1678

For detecting PNPLA3/Pnpla3/Adpn based on the principle of competitive Enzyme Immunoassay

Specifications

Calibration Range 0.1 to 1000ng/mL
Conjugate Biotin
Cross Reactivity Human, Mouse, Rat
Description PNPLA3/Pnpla3/Adpn (Human, Mouse, Rat) ELISA Kit is an in vitro quantitative assay for detecting PNPLA3/Pnpla3/Adpn based on the principle of Competitive Enzyme Immunoassay.
Detection Limit 21pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials Detected Polyclonal Antibody, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials Biotinylated peptide, 8μL HRP Streptavidian concentrate, 100
Quantity 1 Kit
Sample Type Cell Lysate, Culture Supernatant, Plasma, Serum
Sample Volume 100 μL
Content And Storage Store Standard, Biotinylated Adiponutrin peptide, and Positive Control at -80°C. Store the other components at -20°C.
For Use With (Application) Quantification
Product Type PNPLA3 (Human/Mouse/Rat) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.