Learn More
Abnova™ Porcine Reproductive and Respiratory Syndrome Virus ELISA Kit
Shop All Abnova Corporation ProductsDescription
Includes
- 96-well plate pre-coated with recombinant PRRSV antigen, 1 plates
- Positive control, 1.6mL
- Negative control, 1.6mL
- Sample diluent buffer, 48mL
- Wash buffer (10x ):Dilution:1:10, 95mL
- HRP conjugated goat anti-porcine PRRSV antibody (RTU), 24mL
- TMB substrate (TMB color developing solution), 24mL
- Stop solution, 12mL
Specifications
Specifications
| Conjugate | HRP |
| Description | Porcine Reproductive and Respiratory Syndrome Virus ELISA Kit is a sandwich enzyme immunoassay for the detection of Porcine Reproductive and Respiratory Syndrome Virus. |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Sample Type | Plasma, Serum |
| Sample Volume | 100 μL |
| Packaging | 96-well |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Qualitative |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.